Imprimer
Clog, shoe and boot rack storage
Fournisseur: TEKNOMEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
CRFS-BRMOB16 CRFS-BRMOB8 CRFS-BRPE12 CRFS-BRPE16 CRFS-BRPE24 CRFS-BRPE36 CRBS-PEDBR8P CRFS-BRPEL24 CRFS-BRWM12 CRFS-BRWM16 CRFS-BRWM36 CRFS-BRWM8 CRFS-SRMO12 CRFS-SRMO24 CRFS-SRMO36 CRFS-SRPE12 CRFS-SRPE24 CRFS-SRPE36 CRFS-SRWM12 CRFS-SRWM24 CRFS-SRWM36
TEKMCRFS-BRMOB16EA 0 EUR
TEKMCRFS-BRMOB16 TEKMCRFS-BRMOB8 TEKMCRFS-BRPE12 TEKMCRFS-BRPE16 TEKMCRFS-BRPE24 TEKMCRFS-BRPE36 TEKMCRBS-PEDBR8P TEKMCRFS-BRPEL24 TEKMCRFS-BRWM12 TEKMCRFS-BRWM16 TEKMCRFS-BRWM36 TEKMCRFS-BRWM8 TEKMCRFS-SRMO12 TEKMCRFS-SRMO24 TEKMCRFS-SRMO36 TEKMCRFS-SRPE12 TEKMCRFS-SRPE24 TEKMCRFS-SRPE36 TEKMCRFS-SRWM12 TEKMCRFS-SRWM24 TEKMCRFS-SRWM36
Clog, shoe and boot rack storage
Mobilier Portemanteaux
Our hygienically designed stainless steel shoe and boot racks come with a hygienic, easier to clean, continuous piece of prongs.
  • Supplied fully welded, ready to install
  • 304 grade stainless steel, brushed dull polish finish
  • Free standing with two stainless steel levelling feet and non-marking rubber base and front
  • Wall mounted option to save space
  • Two fully welded stainless steel plates at the rear
  • Mobile version also available!
With a narrower and taller design, it provides enough space so they can be thoroughly cleaned on the rack. Choose from bootEn savoir plus