Imprimer
KLEEN blade management system
Fournisseur: SWANN MORTON
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5701 5703 5705 5706 5707 5708 5710 5711 5791 5792 5606
SWAN5701EA 274 EUR
SWAN5701 SWAN5703 SWAN5705 SWAN5706 SWAN5707 SWAN5708 SWAN5710 SWAN5711 SWAN5791 SWAN5792 SWAN5606
KLEEN blade management system
Rasoirs et lames
The KLEEN Blade Management System allows for effortless safe attachment and removal of surgical blades from a standard handle. The system which incorporates a quality Swann-Morton surgical blade is designed to operate with most No.3 and No.4 standard surgical handles already in circulation within the hospital system thus minimising disruption and cost during the conversion process.
  • Sterile and non sterile blades
  • Protective sliding sheath
The sheath covers the blade completely thus assisting with accident free handling. It fully exposes the surgicalEn savoir plus
New Product
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable