Imprimer
Cole-Parmer® Stuart WS-200 Cabinet-Style Water Stills
Fournisseur: Cole Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
99294-05 99294-10 99294-20 99294-25 99294-00 99294-15
STUA99294-05EA 5690 EUR
STUA99294-05 STUA99294-10 STUA99294-20 STUA99294-25 STUA99294-00 STUA99294-15
Cole-Parmer® Stuart WS-200 Cabinet-Style Water Stills
Purificateurs d'eau Distillateurs
Produces cold distilled water for immediate use.
  • Provides highest purity from virtually any raw water supply
  • Reservoir water level sensor turns still off when reservoir is full
  • Simple conversion to pretreated feed
  • Safety features allow for unattended operation
  • Operating pressure: 0,2 - 6,9 bar
Remove organics, bacteria, and pyrogens from your water with the WS-200 cabinet-style water stills. Borosilicate glassware coupled with silica-sheathed heaters enclosed in a protective cabinet, deliver pyrogen-free distillate of the highest purity from virtuallyEn savoir plus