Imprimer
Environmental Express® HotBlock® 200 Digestion Systems
Fournisseur: Cole Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
53206-00 53206-03
STUA53206-00EA 0 EUR
STUA53206-00 STUA53206-03
Environmental Express® HotBlock® 200 Digestion Systems
Systèmes de digestion Systèmes de minéralisation
Easy-to-use external controllers feature automatic shut-off, alarms,and ability to save profi les with ramp and soak settings. Order heating block and control.
  • Eliminate corrosion and sample contamination made of nonmetallic materials
  • Easily transport samples with included polycarbonate transfer racks
  • Mount external controller safely outside the hood