Imprimer
Cole-Parmer® Digital Water Baths
Fournisseur: Cole Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
16340-65
STUA16340-65EA 1270 EUR
STUA16340-65
Cole-Parmer® Digital Water Baths
Bains Bains-marie
Pair with Stuart rotary evaporators for maximum efficiency and smallest footprint.
  • Precise digital temperature display
  • Corrosion-resistant bowl
  • Easy to clean
  • Integrated carry handle for water bath
  • Cool-touch outer case
The clear LED digital display shows the actual bath temperature or the set temperature. There are also two water bath options. The 90 °C bath has a 304 stainless steel bowl and is for water only, suitable for low boiling point applications. The higher temperature 180 °C bath has an aluminium bowl with PTFE coating and can be used withEn savoir plus