Imprimer
Cole-Parmer® Stuart CC-200 Digital Colony Counters
Fournisseur: Cole Parmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
14212-04 14212-06
STUA14212-04EA 1250 EUR
STUA14212-04 STUA14212-06
Cole-Parmer® Stuart CC-200 Digital Colony Counters
Compteurs de colonies
Versatile unit can be used with 50 to 90 mm Petri dishes.
  • Compatible with felt tip markers for pressure-sensitive counting
  • Read individual plates or calculate an average over multiple plates
  • Energy-saving bright-white, LED light source
  • Antimicrobial coating provides additional protection
  • Audible count confirmation, adjustable pressure sensitivity, and option for light/dark background increases flexibility
  • Connects to a printer or computer to record individual or average plate counts
This colony counter registers a count each time you press a felt tipEn savoir plus