Imprimer
EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kits
Fournisseur: STEMCELL Technologies
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
17871 17871RF
STMC17871EA 1450 EUR
STMC17871 STMC17871RF
EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kits
Milieux de séparation cellulaire
Column-free immunomagnetic positive selection of human CD3+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
  • Compatible across EasySep™ "The Big Easy", EasyEights™, and RoboSep™-S platforms
The EasySep™ HLA Chimerism Whole Blood CD3 Positive Selection Kit is designed to isolate CD3+ cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD3 and dextran-coated magnetic particles. The cocktail also contains an antibody to human FcEn savoir plus