Imprimer

Iron(II) sulphate heptahydrate ≥99.5%, granuleux FCC

Marques : Spectrum Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): Iron vitriol, Ferrous sulphate heptahydrate

F1062-2.5KG F1062-500GM
SPCMF1062-2.5KGEA 548 EUR
SPCMF1062-2.5KG SPCMF1062-500GM
Iron(II) sulphate heptahydrate ≥99.5%, granuleux FCC
Fer (II) sulfate heptahydraté

Ferrous Sulfate, Heptahydrate, Granular, FCC is used as an iron fortification food supplement additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark.

Formule: FeSO₄·7H₂O
Poids moléculaire: 278.02 g/mol
Point de fusion: 64
Température de stockage: Température ambiante
Numéro MDL: MFCD00149719
Numéro CAS: 7782-63-0
Indice Merck: 13,04091

Commander maintenant

Résultats des tests de spécification

Assay (FeSO₄.7H₂O) 99.5 - 104.5%
Lead (Pb) max. 2 mg/kg
Mercury max. 1 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR