Imprimer

Acetic acid glacial_AcOH 99,5 USP

Marques : Spectrum Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Acide éthanoïque

AC110-100ML
SPCMAC110-100MLEA 89.1 EUR
SPCMAC110-100ML
Acetic acid glacial_AcOH 99,5 USP
Acide acétique

Glacial Acetic Acid, USP does have some antibacterial and antifungal properties but is used primarily as a pH adjuster. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formule: H₃CCOOH
Poids moléculaire: 60.05 g/mol
Point d'ébullition: 118 °C (1013 hPa)
Point de fusion: 17 °C
Densité: 1,049
Point éclair: 38,5 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00036152
Numéro CAS: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8.3 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR