Imprimer
Oligo(dT)₁₈ primer
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SO132 SO131
SO132EA 102 EUR
SO132 SO131
Oligo(dT)₁₈ primer
Réactifs pour acides nucléiques Primers and Probes
Oligo(dT)₁₈ primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready to use, 20X concentrated aqueous solution. Oligo(dT)₁₈ primer has been functionally tested in first strand cDNA synthesis.