Imprimer
Water Disinfector, WD6010
Fournisseur: SMEG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
WD6010
SMEGWD6010EA 27460 EUR
SMEGWD6010
Water Disinfector, WD6010
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
WD6010 is a single or double glass drop down door washer-disinfector. washing capacity of up to 12 din baskets with 65 cm width. the drop down door allows the operator to ergonomically load and unload the baskets included in the trolley without having to lift heavy weights. also available in a version with turbo cycles (WD6010T model).