Imprimer
Laboratory Glassware Washers, Without Drying System
Fournisseur: SMEG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
GW0260-00-00J GW1260-SC-000 GW1260-S0-000 GW1260-00-000 GW0260-S0-00J
SMEGGW0260-00-00JEA 5940 EUR
SMEGGW0260-00-00J SMEGGW1260-SC-000 SMEGGW1260-S0-000 SMEGGW1260-00-000 SMEGGW0260-S0-00J
Laboratory Glassware Washers, Without Drying System
Lave-verres
The color touch display enables simple and intuitive daily operation. The compact design of the interface ensures the operator a full range of information, including specific programs for every need. In addition, thanks to the ECO program, it is possible to activate a washing cycle with a lower water and energy impact than traditional cycles.
  • Single swing door in stainless steel
USB input enables convenient and efficient laboratory data recording, as well as immediate connection to PC via software for diagnostics, cycle customisation and timely dataEn savoir plus