Imprimer
DNA ladder, O'RangeRuler™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SM0613 SM0623 SM0633 SM0643 SM0653 SM1323 SM1313 SM1303
SM0613EA 135 EUR
SM0613 SM0623 SM0633 SM0643 SM0653 SM1323 SM1313 SM1303
DNA ladder, O'RangeRuler™
Réactifs d'électrophorèse Marqueurs de poids moléculaires
O'RangeRuler™ DNA ladders are step ladders that consist of basic unit repeats of 10, 15, 20, 50, 100, 200, or 500 bp. The ladders have a higher density of bands at a specific DNA size range and are therefore suitable for more accurate sizing of DNA in agarose or non denaturing polyacrylamide gels.
  • Step ladders - ligation products of DNA fragments of equal increments
  • Sharp bands, evenly spaced reference bands
  • Ready to use - premixed with loading dye for direct loading
  • Room temperature storage
  • Supplied with loading dye for sample DNA
Informations de livraison: Supplied with 6X orange DNA loading dye for sample DNA.