Imprimer
Dry swab with transport tube, Q-SWAB™
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
T307-10A T307-5A
SIMPT307-10AEA 50.3 EUR
SIMPT307-10A SIMPT307-5A
Dry swab with transport tube, Q-SWAB™
Écouvillons Écouvillons en tubes
Sterile specimen collection swab.
  • Polypropylene tubes are translucent
  • Cap equipped with silicone washer
  • Non graduated
Q-Swab™ is self-standing, but features a round bottom. The polyester swab holds securely inside the screw cap, making it ideal for specimen collection and protection from contamination. The polypropylene tubes are translucent, making it easy to see through. The cap features an exclusive silicone washer fitted inside to ensure a positive seal at any temperature.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'En savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable