Imprimer
Prismatic film applicators
Fournisseur: Industrial Physics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
VF2163 VF2162 VF2161 VF1837 VF1535 VF1530 VF2164 VF1536 VF1580 VF1581
SIMEVF2163EA 897 EUR
SIMEVF2163 SIMEVF2162 SIMEVF2161 SIMEVF1837 SIMEVF1535 SIMEVF1530 SIMEVF2164 SIMEVF1536 SIMEVF1580 SIMEVF1581
Prismatic film applicators
Applicateurs de films Applicateurs de film pour peintures
Prismatic film applicator with four application sides for applying paint films of four different predefined thicknesses. The prism applicators are available in film widths of 50, 75, 80 and 100 mm and are suitable for applying a host of different products onto flat and relatively solid substrates. Made from high grade stainless steel, the prismatic film applicator will not be affected by acid or base elements.