Imprimer

Rappaport Vassiliadis broth, modified, suitable for microbiology, NutriSelect® Plus

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

17173-500G
SIAL17173-500GEA 120 EUR
SIAL17173-500G
Rappaport Vassiliadis broth, modified, suitable for microbiology, NutriSelect® Plus
Milieux pour microbiologie Milieux pour microbiologie déshydratés

Used as a selective enrichment medium for the isolation of Salmonella species from food and environmental specimens.


  • pH: 5,2 ±0,2 (25 °C)
  • Limited shelf life


Contains papaic digest of soyabean meal 5 g/1110 ml; Sodium chloride 8 g/1110 ml; Monopotassium phosphate 1,6 g/1110 ml; Magnesium chloride anhydrous 18,7 g/L (is equivalent to 40 g/1110 ml Magnesium chloride hexahydrate) and malachite green 0,04 g/1110 ml.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR