Imprimer
SwitchHolder
Fournisseur: SenseAnywhere
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0630125977521
SENA0630125977521EA 15 EUR
SENA0630125977521
SwitchHolder
Enregistreurs de données
The SenseAnywhere SwitchHolder is an accessory which enables you to disable the programmed alarm function of your AiroSensor with a single sliding movement. Just insert your AiroSensor and slide to one side to either activate or disable the alarm notifications of that specific device. This accessory is ideal for environments with cooling systems that are not operational during specific times, i.e. nights or weekends. The SwitchHolder is compatible with the AiroSensor and AiroSensor T. Comes with different labels so you can choose the symbols that match the functionality of your SwitchHolderEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Interface