Imprimer
Culture tubes with screw cap
Fournisseur: schuett-biotec GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3563423 3560103 3562123 3562143 3563143 3563523 3561103
391-0157EA 228 EUR
391-0157 391-6135 SCUT3562123 SCUT3562143 SCUT3563143 SCUT3563523 SCUT3561103
Culture tubes with screw cap
Tubes Tubes de culture
Tubes made of borosilicate with aluminium or plastic screw caps and rubber seal.
  • Heavy walled
  • Easy to inoculate and clean