Imprimer
Loop steriliser, Solaris
Fournisseur: schuett-biotec GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3399002
SCUT3399002EA 2380 EUR
SCUT3399002
Loop steriliser, Solaris
Stérilisateurs Stérilisateurs pour aiguilles
The Schuett Solaris is the perfect solution to avoid using naked flames or if gas cartridges are not available. The unit uses standard electrical power and reaches the safe working temperature of 900 to 1300 °C within seconds. No warm-up time, no excess heat, no energy consumption between work cycles. Start is generated by an infra-red sensor, which allows efficient one-handed operation. The integrated timer can be set to 5 or 7 s. Pathogenic material is kept inside the sterilisation chamber. No spreading, no sooting. Ideal for use in Laminar Flow Cabinets. Working angle can be adjusted.En savoir plus