Imprimer
4-way collector (the werner)
Fournisseur: S.C.A.T.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
106660
SCTV106660EA 312 EUR
SCTV106660
4-way collector (the werner)
Collecteurs
This purging manifold alows you to install up to four SafetyCaps on your purging bottle.
  • Safe and easy handling - capillaries are reliably fixed
  • Solvent filters have enough space due to the wide GLS 80 opening
  • Simply close unused connections with standard bottle closures
The System for Solvent Safety stays permanently closed, the 'safety chain' remains intact during the purging process.