Imprimer
Ballons d'évaporation, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
241202707 241203703 241204605 241205601
SCOT241202707EA 49.6 EUR
SCOT241202707 SCOT241203703 SCOT241204605 SCOT241205601
Ballons d'évaporation, DURAN®
Ballons Flacons pour évaporateurs rotatifs
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
NS 29/32 standard ground joint socket, pear-shaped, centre neck, thick-walled. Suitable for all standard rotary evaporators; with white labelling area.