Imprimer
Redox combination electrodes, AquaLine
Fournisseur: SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
285113073
SCOI285113073EA 228 EUR
SCOI285113073
Redox combination electrodes, AquaLine
Électrodes Électrodes redox
AquaLine electrodes are ideal for measuring applications in drinking water, industrial water and sewage range.