Imprimer
Darkfield optic ring for VisiLED slim ring light
Fournisseur: SCHOTT AG Lighting and imaging
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
400810
SCOC400810EA 275 EUR
SCOC400810
Darkfield optic ring for VisiLED slim ring light
Éclairages Éclairages annulaires
Focus optic ring for darkfield illumination.
  • Focus optics ring for darkfield
  • Working distance: 5 to 25 mm
Designed for use only with VisiLED slim ring light (SCOC400150).
Informations de commande: VisiLED slim ring light and controller MC 750, MC 1100, or MC 1500 required.