Imprimer
Culture shake flasks, with baffles, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SP25630-1000 SP25630-2000 SP25630-500 SP25630-250
SCERSP25630-1000EA 139 EUR
SCERSP25630-1000 SCERSP25630-2000 SCERSP25630-500 SCERSP25630-250
Culture shake flasks, with baffles, KIMBLE®
Ballons Flacons de culture
These baffled culture shake flasks are designed for use with rotary and reciprocating shakers where baffle indents provide greater turbulence to improve oxygen transfer rates or aeration.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • All flasks have durable white ceramic enamel marking spots
  • Flask has a neck with a plain fire-polished finish and suitable for a metal cap, plastic cap, foam plug or cotton plug
  • Flasks feature 3 baffles