Imprimer
ThermoSafe® PolarPack® Gel Packs
Fournisseur: Sonoco Thermosafe
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
PP48 PP6 PP8BLANKFILM
SCATPP48EA 33.8 EUR
SCATPP48 SCATPP6 SCATPP8BLANKFILM
ThermoSafe® PolarPack® Gel Packs
Poches refroidissantes
PolarPack® gel packs provide long lasting, high performance temperature protection.
  • Gel filled pouch
  • Can be refrigerated or frozen to keep products cold or frozen
  • Suitable for pharmaceuticals, clinical trials, diagnostic kits, chemicals, and biologicals
  • Available in a range of sizes
These packs are designed to safeguard temperature-sensitive products during storage and transport. They can be chilled in a freezer, refrigerator, or low temperature incubator and are effective when used with insulated shipping containers. Phase changes at 0 °C and returnsEn savoir plus