Imprimer
Tork W14 Hand Cleaning Wet Wipes, Refill
Fournisseur: Essity
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
190692
SCAA190692EA 77.8 EUR
SCAA190692
Tork W14 Hand Cleaning Wet Wipes, Refill
Lingettes Lingettes et essuie-mains à usage unique Lingettes présaturées
Clean hands effectively with these professional-quality wipes.
  • Remove oil, grease, lubricants and dirt with ease, thanks to the strong texture and high absorbency of these wet wipes
  • Avoid potential irritations with wipes that are free of fragrance and kind to the skin
  • Refilling is easy and efficient with the bucket-style dispenser
When there is no access to water, these Tork hand cleaning wet wipes can be used. Their strong texture works to swiftly remove oil, grease and lubricants from hands. It can tackle dirt effectively, too, thanks to theirEn savoir plus