Imprimer
Prefilter attachment
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
16807
SART16807EA 282 EUR
SART16807
Prefilter attachment
Porte-filtres
Allows the separation of coarse solids from samples with bacteriological germ detection in a single operation, before and during the actual germ-retaining filtration step.