Imprimer
CO₂ incubators, MCO series, with safecell UV®, PHCbi
Fournisseur: PHC Corporation
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MCO-230AICUVPE
SANYMCO-230AICUVPEEA 13060 EUR
SANYMCO-230AICUVPE
CO₂ incubators, MCO series, with safecell UV®, PHCbi
Incubateurs Incubateurs CO₂
These incubators offer quality in performance, maximise cell culture productivity and provide optimum results and reproducibility.
  • Improved insulation performance and lower running costs
  • PID Control of CO₂ and temperature
  • Dual IR CO₂ sensor
  • Optimum protection and space for cell culturers
  • Easy to operate and maintain
They provide precisely controlled environment for sensitive cell cultures, delivering long-term performance adn optimal cell viability. Also they support a reliable, stable cell culture environment across all shelf positions, each andEn savoir plus