Imprimer
Beakers with Spout (316L Stainless Steel)
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
A866-100 A866-1000 A866-200 A866-2000 A866-3000 A866-500 A866-5000
SAMPA866-100EA 212 EUR
SAMPA866-100 SAMPA866-1000 SAMPA866-200 SAMPA866-2000 SAMPA866-3000 SAMPA866-500 SAMPA866-5000
Beakers with Spout (316L Stainless Steel)
Béchers
These beakers with spout are excellent for handling and dispensing a variety of liquids.
  • Made from high grade 316L stainless steel
  • Excellent corrosion resistance
  • Wide range of sizes available
These stainless steel beakers with spout are made from top quality 316L stainless steel which provides excellent corrosion resistance and provides an ideal way to store, handle and dispense a wide variety of liquids across many industries.