Imprimer
Cup samplers
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
5030A-1500 5030A-1000 5030A-600 5030J-1000 5030J-1500
SAMP5030A-1500EA 1010 EUR
SAMP5030A-1500 SAMP5030A-1000 SAMP5030A-600 SAMP5030J-1000 SAMP5030J-1500
Cup samplers
Échantillonneurs Préleveurs de liquides
Stainless steel cup samplers designed for liquids and suspensions.
  • Single point liquid sampler with removable cup
  • Simple operation, easy to clean and hygienic design
  • Discrete samples to be taken from known heights within the product