Imprimer
Goat serum
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
S2000-100 S2000-500 S200H-500 S200H-100
S2000-100EA 30 EUR
S2000-100 S2000-500 S200H-500 S200H-100
Goat serum
Sérums
Goat serum is a blood derived product collected from healthy non immunised animals.
  • Blocking agent or negative control in immunoassays
  • For immunohistochemistry and immunocytochemistry, used between 1 to 20% as a blocking agent in various studies
Suitable alternative to FBS in a wide range of cell culture applications (e.g. isolation and identification of viruses produced in a variety of primary cells).