Imprimer
Beaker tongs, stainless steel/rubber
Fournisseur: RSG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
043.290
RSGA043.290EA 36.3 EUR
RSGA043.290
Beaker tongs, stainless steel/rubber
Pinces
Stainless steel 18/10. All-purpose beaker tongs with rubber protected tips. With second diameter for holding small flasks and beakers.