Imprimer
Prohibition signs, Sekuroka®
Fournisseur: Roth Carl
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
X450.1 0206.1 0207.1 0216.1 0462.1
ROTHX450.1EA 16.2 EUR
ROTHX450.1 ROTH0206.1 ROTH0207.1 ROTH0216.1 ROTH0462.1
Prohibition signs, Sekuroka®
Panneaux
Made of self-adhesive PVC foil, 0,1 mm, with permanent-glue and colour-fast print. Ideal for indoor and outdoor use.
Certifications: According to DIN 4844, BGV A8 and field-proven safety marking.