Imprimer

Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fournisseur: Rockland Immunochemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

613-4204
ROCK613-4204EA 324 EUR
ROCK613-4204
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Secondary Rabbit Anti-F(ab')2 Reacts with Sheep (Ovine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR