Imprimer

Anti-IgG (H+L) Sheep Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Fournisseur: Rockland Immunochemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

612-6002
ROCK612-6002EA 248 EUR
ROCK612-6002
Anti-IgG (H+L) Sheep Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anticorps
Secondary Sheep Anti-IgG (H&L) Reacts with Rat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Sheep
Isotype: IgG
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR