Imprimer

Anti-IgG (H+L) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Fournisseur: Rockland Immunochemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

612-1025
ROCK612-1025EA 299 EUR
ROCK612-1025
Anti-IgG (H+L) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anticorps
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: Polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR