Imprimer

Anti-GYS1 Rabbit Polyclonal Antibody

Fournisseur: Rockland Immunochemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

600-401-B28
ROCK600-401-B28EA 1180 EUR
ROCK600-401-B28
Anti-GYS1 Rabbit Polyclonal Antibody
Anticorps
This phospho specific polyclonal antibody was tested by immunoblotting and ELISA.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: GYS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR