Imprimer

Anti-KCND3 Mouse Monoclonal Antibody (Unconjugated) [clone: S75-41]

Fournisseur: Rockland Immunochemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

200-301-G04
ROCK200-301-G04EA 841 EUR
ROCK200-301-G04
Anti-KCND3 Mouse Monoclonal Antibody (Unconjugated) [clone: S75-41]
Anticorps
Anti-Kv4.3 Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~75kDa in Rat brain membrane preparations. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: KCND3
Clonality: Monoclonal
Clone: S75-41
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Xenopus, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR