Imprimer

Anti-FITC Goat Polyclonal Antibody

Fournisseur: Rockland Immunochemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

100-1196
ROCK100-1196EA 743 EUR
ROCK100-1196
Anti-FITC Goat Polyclonal Antibody
Anticorps
Anti-Fluorescein Antibody has been assayed against 1.0 µg of Fluorescein conjugated Rabbit IgG in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Donkey)5 and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: pan
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR