Imprimer
Filter discs, biplane, series 16, porosity 4, VitraPOR®
Fournisseur: ROBU GLASFILTER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
16104 16304 16404 16504 16604 16704 16804 16904 16114 16204 16254 16124 62004 61904 62304 62704 62604 62804 62504 62404 63804 63004 62904 63504 61504 61704 61604 61804 61404 61304 62204 62104
ROBU16104EA 85.3 EUR
ROBU16104 ROBU16304 ROBU16404 ROBU16504 ROBU16604 ROBU16704 ROBU16804 ROBU16904 ROBU16114 ROBU16204 ROBU16254 ROBU16124 ROBU62004 ROBU61904 ROBU62304 ROBU62704 ROBU62604 ROBU62804 ROBU62504 ROBU62404 ROBU63804 ROBU63004 ROBU62904 ROBU63504 ROBU61504 ROBU61704 ROBU61604 ROBU61804 ROBU61404 ROBU61304 ROBU62204 ROBU62104
Filter discs, biplane, series 16, porosity 4, VitraPOR®
Filtres Disques filtrants
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.