Imprimer
Vacuum-/gas distribution filters, VitraPOR®
Fournisseur: ROBU GLASFILTER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
18500 18501 18502 18503 18504
ROBU18500EA 46.4 EUR
ROBU18500 ROBU18501 ROBU18502 ROBU18503 ROBU18504
Vacuum-/gas distribution filters, VitraPOR®
Filtres Filtres de traitement des liquides
These filters are suitable for degassing and filtration of solvents e.g. in chromatography.
  • Good chemical resistance
  • Good temperature resistance
Made of borosilicate glass.