Imprimer
Filter candles, cylindrical, VitraPOR®
Fournisseur: ROBU GLASFILTER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
18403 18404 18410 18411 18412 18413 18414 18402 18340 18341 18342 18343 18344 18350 18351 18352 18353 18354 18400 18401
ROBU18403EA 211 EUR
ROBU18403 ROBU18404 ROBU18410 ROBU18411 ROBU18412 ROBU18413 ROBU18414 ROBU18402 ROBU18340 ROBU18341 ROBU18342 ROBU18343 ROBU18344 ROBU18350 ROBU18351 ROBU18352 ROBU18353 ROBU18354 ROBU18400 ROBU18401
Filter candles, cylindrical, VitraPOR®
Filtres Tubes filtrants
These cylindrical filter candles are available with or without tube.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
Made of borosilicate glass 3.3.