Imprimer
Foot adapter
Fournisseur: R L ENTERPRISES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2119
RLAA2119EA 27.4 EUR
RLAA2119
Foot adapter
Pinces Noix de serrage
Adapter manufactured out of aluminium with boss to fit scaffold feet.
Accepts three 13 mm diameter rods or tubes at right angles (2 perpendicular to the boss and the third in line with the boss), with alien screws for fixing. One allen key provided per pack of ten.