Imprimer
Reticulocyte stain, Sigma-Aldrich®
Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
R4132-120ML
R4132-120MLEA 165 EUR
R4132-120ML
Reticulocyte stain, Sigma-Aldrich®
Reticulocyte stain
Utilizes New Methylene Blue for the identification of reticulocytes in blood films.
IVDLogo