Imprimer

Anti-H1H3592P3 Recombinant Antibody

Fournisseur: ProSci Inc.
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

10-776
PRSI10-776EA 654 EUR
PRSI10-776
Anti-H1H3592P3 Recombinant Antibody
Anticorps
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR