Imprimer
TCH defined serum-free growth medium
Marques : PROTIDE PHARMACEUTICALS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3001 3002 3005
PRPH3001EA 0 EUR
PRPH3001 PRPH3002 PRPH3005
TCH defined serum-free growth medium
Suppléments et additifs pour culture cellulaire
Serum replacement used primarily for human cells and the production of cell secreted proteins. TCH™ is also effective in culturing other mammalian cells without the use of animal proteins. Contains only human proteins (i.e., human serum albumin, human transferrin, recombinant insulin), which are heat treated.
  • Defined, serum-free, xeno-free
  • No hormones, antibiotics, growth factors, phenol red, Ig
  • 50X concentrate
  • Can be utilised with a wide variety of basal media
Certifications: cGMP manufactured.