Imprimer
Ion chromatography standard, lithium, VHG
Marques : LGC Standards PROMOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
VHG-ILI100-500 VHG-ILIW1K-100
PROOVHG-ILI100-500EA 0 EUR
PROOVHG-ILI100-500 PROOVHG-ILIW1K-100
Ion chromatography standard, lithium, VHG
Étalons Étalons cationiques Standards de lithium
This solution is intended for use as a certified reference material and calibration standard, for ion chromatography (IC) and techniques using other modes of aqueous ion detection.
Certifications: Traceable to NIST and SRM.