Imprimer
Clino Mopping System
Fournisseur: PFENNIG REINIGUNGSTECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2009000 2500050 2009500
PPSA2009000EA 64.5 EUR
PPSA2009000 PPSA2500050 PPSA2009500
Clino Mopping System
Serpillières
Mop frame is suitable for use with all Pfennig mop heads: standard, professional, and universal.
  • Sturdy and durable stainless steel construction
  • Sterilizable
  • Contact-free mop drop
  • Also available with locking-screw
Informations de commande: Mop frames and handles are sold separately. Telescopic alumimium mop handle must be ordered along with a mop frame to complete the system.