Imprimer

Desalting columns, Zeba™, Pierce™

Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

89877 89882 89890 89894 89891 89893 89889 89892 89878 89883 87773
PIER89877EA 200 EUR
PIER89877 PIER89882 PIER89890 PIER89894 PIER89891 PIER89893 PIER89889 PIER89892 PIER89878 PIER89883 PIER87773
Desalting columns, Zeba™, Pierce™
Colonnes pour chromatographie
These spin desalting columns are made of low protein binding PP and are compatible with a wide range of standard laboratory instruments and consumables. They provide >95% retention of salts and other small molecules.

  • Higher protein recovery
  • Minimal sample dilution
  • No screening fractions for protein or waiting for protein to emerge by gravity-flow
  • Easy-to-use with no cumbersome column preparation or equilibration

The Zeba 7K MWCO is recommended for processing proteins >7 kDa, and the Zeba 40K MWCO for processing proteins >40 kDa.

Attention:En savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR