Imprimer
Inclusion body solubilisation reagent, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
78115
PIER78115EA 293 EUR
PIER78115
Inclusion body solubilisation reagent, Pierce™
Protéines purification Réactifs de purification des protéines
Inclusion Body Solubilisation Reagent is designed to retrieve expressed protein in soluble form after lysis and extraction with reagents.
  • Achieves the essential first step for subsequent insoluble protein refolding procedures
  • One easy to use solution
  • Proprietary denaturant can be easily removed by dialysis so that SDS-PAGE may be performed
  • Can accommodate additional components such as reducing agents or chelating agents